Learn More
Medchemexpress LLC HY-136605 5mg , DGY-06-116 CAS:2556836-50-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1366055MG
Encompass_Preferred
HY-136605 5mg DGY-06-116 CAS: 2556836-50-9 DGY-06-116 is an irreversible covalent, selective Src inhibitor with an IC50 of 3nM. DGY-06-116 inhibits FGFR1 with an IC50 of 8340 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0088