Learn More
AMID BIOSCIENCES Amid Biosciences, DH10B Chemically Competent E. coli Cells, Large DNA Cloning, 10 x 0.1 mL
SDP
Supplier:  AMID BIOSCIENCES DH10B201
Encompass_Preferred
DH10B chemically competent E. coli cells are supplied in a 10 x 0.1 mL format for high quality cloning and propagation of large or complex DNA constructs. DH10B is valued for stable plasmid maintenance and molecular cloning workflows where insert integrity and transformation performance matter. Applications include large DNA cloning, stable propagation of large plasmids, cloning workflows requiring high-quality transformation host. Technical specifications are included below.- Quantity - 10 x 0.1 mL/tube- Organism - E. coli- Storage - -80 C- Cell Strain / Host - DH10B- Transformation Efficiency - 1.00E+9- Cell Format - 10 x 0.1 mL chemically competent cellsThis product is for research use only.
Catalog No. NC4043488