Learn More
Gold Biotechnology Inc GB10B-Pro™ Electrocompetent E. coli Cells 5x50 µl
SDP
Numéro de catalogue. NC2469185
Encompass
GoldBio’s GB10B-Pro™ Electrocompetent E. coli cells are especially designed for the most demanding cloning applications. GB10B-Pro™ cells will provide the greatest number of transformants for when your research requires it, including assembling large and multi-DNA fragments, cloning large (≥10 kb up to 350 kb) or difficult construct transformations, working with synthetic bio-applications, and even BAC cloning.
Numéro de catalogue. NC2469185 Fournisseur Gold Biotechnology Inc Code fournisseur CC2015X50
Ajouter au panier
Ajouter au panier
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
GoldBio’s GB10B-Pro™ Electrocompetent E. coli cells are especially designed for the most demanding cloning applications. GB10B-Pro™ cells will provide the greatest number of transformants for when your research requires it, including assembling large and multi-DNA fragments, cloning large (≥10 kb up to 350 kb) or difficult construct transformations, working with synthetic bio-applications, and even BAC cloning.
missing translation for 'productTitle'
missing translation for 'pleaseSelectIssue'
missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.