Learn More
Medchemexpress LLC DHA ceramide | 218608-43-6 | 98.7% | 609.96 | C40H67NO3 | 10 MG
SDP
Catalog No. 5000289289
Encompass_Preferred
DHA Ceramide (C22:6 Ceramide) is a high-purity ester-form ceramide supplied as a solid for biochemical and cell biology research. It is intended for in vitro use and is typically prepared in DMSO for cellular treatments. Store the dry material at -20°C protected from light and under nitrogen; reconstituted solutions have recommended cold storage lifetimes.
  • High purity (98.65%) suitable for research applications.
  • Available in small milligram-scale packages for flexible experimental use.
  • Solid form soluble in DMSO at 100 mg/mL; ultrasonic treatment recommended.
  • Provides molecular formula C40H67NO3 and molecular weight 609.96 g/mol for experimental planning.
  • Recommended storage: store at -20°C protected from light and under nitrogen; solutions: -80°C (6 months) or -20°C (1 month).
Catalog No. 50-002-89289 Supplier Medchemexpress LLC Supplier No. HY16578110MG
DHA Ceramide (C22:6 Ceramide) is a high-purity ester-form ceramide supplied as a solid for biochemical and cell biology research. It is intended for in vitro use and is typically prepared in DMSO for cellular treatments. Store the dry material at -20°C protected from light and under nitrogen; reconstituted solutions have recommended cold storage lifetimes.
  • High purity (98.65%) suitable for research applications.
  • Available in small milligram-scale packages for flexible experimental use.
  • Solid form soluble in DMSO at 100 mg/mL; ultrasonic treatment recommended.
  • Provides molecular formula C40H67NO3 and molecular weight 609.96 g/mol for experimental planning.
  • Recommended storage: store at -20°C protected from light and under nitrogen; solutions: -80°C (6 months) or -20°C (1 month).