Learn More
HORIZON DISCOVERY BIOSCIENCES LIMITED Dharmacon Accell siRNA, Human VCAM1, Chemically Modified, 4/Pk.
Catalog No. NC3924697
Encompass
This product consists of a set of four annealed, double-stranded RNA oligonucleotides designed for RNA interference targeting VCAM1. These reagents feature chemical modifications that enhance stability, target specificity, and cellular uptake in the absence of exogenous transfection reagents.
  • Confirmed by MALDI-TOF mass spectrometry for strand mass accuracy
  • Features 3'-UU overhangs and 5'-phosphate on the antisense strand
  • Supports at least 75% gene silencing efficiency
  • Recommended working concentration of 1 µM
  • Optimized mRNA silencing typically achieved by 72 hours
  • Delivery performance sensitive to serum concentration; optimization in serum-free media is recommended
Catalog No. NC3924697 Supplier HORIZON DISCOVERY BIOSCIENCES LIMITED Supplier No. EQ013351000020
This product consists of a set of four annealed, double-stranded RNA oligonucleotides designed for RNA interference targeting VCAM1. These reagents feature chemical modifications that enhance stability, target specificity, and cellular uptake in the absence of exogenous transfection reagents.
  • Confirmed by MALDI-TOF mass spectrometry for strand mass accuracy
  • Features 3'-UU overhangs and 5'-phosphate on the antisense strand
  • Supports at least 75% gene silencing efficiency
  • Recommended working concentration of 1 µM
  • Optimized mRNA silencing typically achieved by 72 hours
  • Delivery performance sensitive to serum concentration; optimization in serum-free media is recommended