Learn More
HORIZON DISCOVERY BIOSCIENCES LIMITED Dharmacon™ ON-TARGETplus 2.0 siRNA, Human, MS4A1, SMARTpool, 10 nmol
Catalog No. NC3939726
Encompass
This product consists of siRNA reagents designed with patented modification patterns and the SMARTselection algorithm to ensure efficient and specific target gene silencing. The chemistry is engineered to destabilize off-target activity by modifying the antisense seed region and preventing non-specific interactions with the RISC complex, while maintaining optimal knockdown performance.
  • Guaranteed gene silencing of at least 75% at the mRNA level when used under optimal delivery conditions
  • Patented modification pattern to reduce off-target effects
  • Sense strand modification to favor antisense strand uptake
  • Antisense strand seed region modification to enhance specificity
  • Design filters to eliminate common seed regions linked to miRNA-like off-targets
  • Available in a pooled format consisting of four siRNAs per reagent
Catalog No. NC3939726 Supplier HORIZON DISCOVERY BIOSCIENCES LIMITED Supplier No. L2012214010010
This product consists of siRNA reagents designed with patented modification patterns and the SMARTselection algorithm to ensure efficient and specific target gene silencing. The chemistry is engineered to destabilize off-target activity by modifying the antisense seed region and preventing non-specific interactions with the RISC complex, while maintaining optimal knockdown performance.
  • Guaranteed gene silencing of at least 75% at the mRNA level when used under optimal delivery conditions
  • Patented modification pattern to reduce off-target effects
  • Sense strand modification to favor antisense strand uptake
  • Antisense strand seed region modification to enhance specificity
  • Design filters to eliminate common seed regions linked to miRNA-like off-targets
  • Available in a pooled format consisting of four siRNAs per reagent