Learn More
HORIZON DISCOVERY BIOSCIENCES LIMITED Dharmacon On-Targetplus siRNA, Human CD38, SMARTpool, 10 nmol
Catalog No. NC3939721
Encompass
These siRNA reagents feature a patented dual-strand chemical modification pattern designed to provide high-specificity gene silencing while significantly reducing off-target activity. By leveraging a sophisticated bioinformatics approach that incorporates seed-region analysis and filtering, these reagents minimize miRNA-like off-targets. The sense strand is modified to inhibit RISC uptake, favoring the antisense strand, while the antisense strand seed region is modified to destabilize off-target interactions.
  • Reduces off-target gene silencing by up to 90%
  • Guaranteed to silence target gene expression by at least 75% at the mRNA level when used under optimal delivery conditions
  • Utilizes a dual-strand modification pattern to address both siRNA strands
  • Incorporates low-frequency seed regions to ensure fewer off-targets
  • Provides sequence information with purchase
Catalog No. NC3939721 Supplier HORIZON DISCOVERY BIOSCIENCES LIMITED Supplier No. L2009222010010
These siRNA reagents feature a patented dual-strand chemical modification pattern designed to provide high-specificity gene silencing while significantly reducing off-target activity. By leveraging a sophisticated bioinformatics approach that incorporates seed-region analysis and filtering, these reagents minimize miRNA-like off-targets. The sense strand is modified to inhibit RISC uptake, favoring the antisense strand, while the antisense strand seed region is modified to destabilize off-target interactions.
  • Reduces off-target gene silencing by up to 90%
  • Guaranteed to silence target gene expression by at least 75% at the mRNA level when used under optimal delivery conditions
  • Utilizes a dual-strand modification pattern to address both siRNA strands
  • Incorporates low-frequency seed regions to ensure fewer off-targets
  • Provides sequence information with purchase