Learn More
HORIZON DISCOVERY BIOSCIENCES LIMITED Dharmacon ON-TARGETplus Synthetic siRNA, Mouse, Enpp2, SMARTpool, 1 ea
Catalog No. NC3914928
Encompass
This synthetic small interfering RNA (siRNA) reagent is designed for high-specificity gene silencing in mouse models. Featuring the SMARTpool format, this product provides a mixture of four distinct siRNA sequences in a single tube to ensure reliable loss-of-function studies and potent knockdown. The reagent incorporates patented chemical modifications that minimize off-target activity and enhance target-specific gene suppression.
  • Guaranteed mRNA knockdown of at least 75%
  • SMARTselection design algorithm for high-confidence silencing
  • Chemically modified strands to prevent nuclease-mediated degradation
  • Antisense strand seed region modifications to destabilize off-target binding
  • Sense strand modifications to favor antisense strand uptake
  • Two RNA strands forming a 21 bp duplex with 3' dinucleotide overhangs
  • Suitable for concentrations of 5 to 25 nM
Catalog No. NC3914928 Supplier HORIZON DISCOVERY BIOSCIENCES LIMITED Supplier No. L048995000020
This synthetic small interfering RNA (siRNA) reagent is designed for high-specificity gene silencing in mouse models. Featuring the SMARTpool format, this product provides a mixture of four distinct siRNA sequences in a single tube to ensure reliable loss-of-function studies and potent knockdown. The reagent incorporates patented chemical modifications that minimize off-target activity and enhance target-specific gene suppression.
  • Guaranteed mRNA knockdown of at least 75%
  • SMARTselection design algorithm for high-confidence silencing
  • Chemically modified strands to prevent nuclease-mediated degradation
  • Antisense strand seed region modifications to destabilize off-target binding
  • Sense strand modifications to favor antisense strand uptake
  • Two RNA strands forming a 21 bp duplex with 3' dinucleotide overhangs
  • Suitable for concentrations of 5 to 25 nM