Learn More
HORIZON DISCOVERY BIOSCIENCES LIMITED DharmaFECT 1 Transfection Reagent | 5x1.5ml
Catalog No. NC1743725
Encompass
The most broadly applicable DharmaFECT formulation for optimal siRNA or microRNA transfection into a wide range of cell types for successful RNAi experiments. DharmaFECT 1 has been validated in over 35 cell types.
Catalog No. NC1743725 Supplier HORIZON DISCOVERY BIOSCIENCES LIMITED Supplier No. T200104
The most broadly applicable DharmaFECT formulation for optimal siRNA or microRNA transfection into a wide range of cell types for successful RNAi experiments. DharmaFECT 1 has been validated in over 35 cell types.