Learn More
HORIZON DISCOVERY BIOSCIENCES LIMITED DharmaFECT 3 Transfection Reagent, 0.75mL
Catalog No. NC1650394
Encompass
One of four siRNA/microRNA specific formulations, DharmaFECT 3 is a chemically distinct alternative to one-size-fits-all transfection reagents to achieve high-efficiency silencing in more cell types.
Catalog No. NC1650394 Supplier HORIZON DISCOVERY BIOSCIENCES LIMITED Supplier No. T200302
One of four siRNA/microRNA specific formulations, DharmaFECT 3 is a chemically distinct alternative to one-size-fits-all transfection reagents to achieve high-efficiency silencing in more cell types.