Learn More
HORIZON DISCOVERY BIOSCIENCES LIMITED DharmaFECT Set of 4 Transfection Reagents, 0.75 mL
Catalog No. NC1585975
Encompass
An aliquot of each of the four DharmaFECT formulations for siRNA/microRNA transfection optimization studies. Useful for determination of the best reagent for your cells and particular experimental conditions.
Catalog No. NC1585975 Supplier HORIZON DISCOVERY BIOSCIENCES LIMITED Supplier No. T200502
An aliquot of each of the four DharmaFECT formulations for siRNA/microRNA transfection optimization studies. Useful for determination of the best reagent for your cells and particular experimental conditions.