Learn More
Selleck Chemical LLC DHBP dibromide S3700-25mg
SDP
Catalog No. 503109282
Encompass_Preferred
DHBP (1 1 -diheptyl-4 4 -bipyridinium) a viologen for electrochromic memory display agent inhibits the calcium release induced by 2 mM caffeine and 2 g/ml polylysine with an IC50 value of 5 g/ml and 4 g/ml respectively
Catalog No. 50-310-9282 Supplier Selleck Chemical LLC Supplier No. S370025MG
DHBP (1 1 -diheptyl-4 4 -bipyridinium) a viologen for electrochromic memory display agent inhibits the calcium release induced by 2 mM caffeine and 2 g/ml polylysine with an IC50 value of 5 g/ml and 4 g/ml respectively