Learn More
Apexbio Technology LLC DHBP dibromide 6159-05-3 100mg
SDP
Catalog No. 5000092306
Encompass_Preferred
DHBP dibromide (CAS 6159-05-3) is a bioactive small molecule that modulates intracellular calcium dynamics by inhibiting calcium release from the sarcoplasmic reticulum Its primary mechanism involves regulating calcium channel activity thereby attenuating cytosolic calcium fluctuations and influencing muscle contraction processes The compound is widely utilized in research exploring excitation-contraction coupling and calcium signaling pathways in skeletal and cardiac muscle Applications include studies in neuroscience muscle physiology and intracellular signal transduction where DHBP dibromide serves as a valuable tool for elucidating molecular events underlying calcium-dependent physiological functions
Catalog No. 50-000-92306 Supplier Apexbio Technology LLC Supplier No. B6468100
DHBP dibromide (CAS 6159-05-3) is a bioactive small molecule that modulates intracellular calcium dynamics by inhibiting calcium release from the sarcoplasmic reticulum Its primary mechanism involves regulating calcium channel activity thereby attenuating cytosolic calcium fluctuations and influencing muscle contraction processes The compound is widely utilized in research exploring excitation-contraction coupling and calcium signaling pathways in skeletal and cardiac muscle Applications include studies in neuroscience muscle physiology and intracellular signal transduction where DHBP dibromide serves as a valuable tool for elucidating molecular events underlying calcium-dependent physiological functions