Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DHC-156 | 99.5% | 492.47 g·mol⁻¹ | C24H24F4N4O3 | 10 MG
SDP

Catalog No. 5000288054
Encompass_Preferred

DHC-156 is a selective small-molecule downmodulator of the transcription factor Brachyury, developed for preclinical research into chordoma. It downmodulates brachyury post-translationally and has been shown to impair chordoma cell growth. Intended for research use only.

  • Selective downmodulator of the transcription factor Brachyury.
  • Downmodulates brachyury post-translationally and impairs chordoma cell growth.
  • Chemical formula: C24H24F4N4O3.
  • Molecular weight: 492.47 g·mol⁻¹.
  • High purity (99.5%).
  • Available in multiple pack sizes, including 10 mg.
  • Supplied for research use only; not for human or clinical use.

Catalog No. 50-002-88054 Supplier Medchemexpress LLC Supplier No. HY15796410MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

DHC-156 is a selective small-molecule downmodulator of the transcription factor Brachyury, developed for preclinical research into chordoma. It downmodulates brachyury post-translationally and has been shown to impair chordoma cell growth. Intended for research use only.

  • Selective downmodulator of the transcription factor Brachyury.
  • Downmodulates brachyury post-translationally and impairs chordoma cell growth.
  • Chemical formula: C24H24F4N4O3.
  • Molecular weight: 492.47 g·mol⁻¹.
  • High purity (99.5%).
  • Available in multiple pack sizes, including 10 mg.
  • Supplied for research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.