Learn More
Enzo Life Sciences Dihydroergocristine . mesylate (200mg). CAS: 24730-10-7
SDP
Supplier:  Enzo Life Sciences BMLNS1080200
Encompass_Preferred
Serotonin antagonist. Dopaminergic and adrenergic partial agonist. Cognition enhancer. Purity: ≥98%. Solubility: Soluble in water (10mg/ml), methanol (22mg/ml), or DMSO (25mM). Long Term Storage: Ambient.
Catalog No. 50-201-1205