Learn More
Enzo Life Sciences DHEA ELISA kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ADI900093
Encompass_Preferred
Highly sensitive ELISA, measuring DHEA in culture supernatants, plasma, serum, saliva, and urine from any species. The DHEA EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 5 hours. Absorbance is read at 405 nm. This kit has low reactivity to structurally related molecules and offers sensitive measurement of DHEA in a variety of sample types. Alternative Name: Dehydroepiandrosterone. Sensitivity: 2.9 pg/ml (range 12.21 - 50,000 pg/ml). Assay Time: 5 hours. Applications: ELISA, Colorimetric detection. Species Reactivity: Species independent. Contents: GxR IgG Microtiter plate, Assay buffer, Conjugate, Antibody, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution
Catalog No. 50-658-138