Learn More
Enzo Life Sciences DHEA ELISA kit (5x96wells)
SDP
Supplier:  Enzo Life Sciences ADI901093
Encompass_Preferred
The DHEA EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 5 hours. Absorbance is read at 405 nm. This kit has low reactivity to structurally related molecules and offers sensitive measurement of DHEA in a variety of sample types. Alternative name: Dehydroepiandrosterone. Assay Time: 5 hours. Applications: ELISA, Colorimetric detection . Species reactivity: Species independent .
Catalog No. 50-200-6894