Learn More
Drg International Inc DRG DHEA-S ELISA, 96 Well Kit
Supplier:  Drg International Inc EIA1562
Encompass_Preferred
An enzyme immunoassay for the quantitative measurement of DHEA-S in serum and plasma. Dehydroepiandrosterone (5-Androstene-3Beta-0L-17-one, Androstenolone, Dehydroisoandrosterone, Transdehydroandrosterone, DHEA) is a steroid hormone present in blood mostly in its sulfate form (DHEA S). The DRG DHEA-S ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA), based on the principle of competitive binding. The microtiter wells are coated with a polyclonal antibody directed towards an antigenic site of the DHEA-S molecule.
Catalog No. 50-187-1232