Learn More
Aviva Systems Biology Corp DHFR2 Antibody - middle region (ARP53341_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53341_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DHFRL1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human DHFRL1. Alternate names: DHFRP4, FLJ16119ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53967