Learn More
Medchemexpress LLC HY-P7338 10ug Medchemexpress, DHH, Human CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP733810UG
Encompass_Preferred
Medchemexpress, HY-P7338 10ug DHH, Human CAS: Recombinant Human Desert Hedgehog produced in E. coli is a single non-glycosylated polypeptide chain. Desert hedgehog protein (DHH) is a member of the Hedgehog family which encodes signaling molecules that play an important role in regulating morphogenesis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0599