Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DHH protein, human (His-GST, Myc) | ≥90.0% | 100UG
SDP

Catalog No. 5000177788
Encompass_Preferred

Recombinant human desert hedgehog (DHH) protein supplied as a lyophilized preparation with N-terminal His and GST tags and a C-terminal Myc tag. Expressed in E. coli, the construct corresponds to amino acids G51-K158 of human DHH (accession O43323) and is intended for research on hedgehog signaling and gonadal development.

  • Recombinant human DHH with His-GST and Myc tags.
  • Expressed in E. coli and lyophilized for stability.
  • Formulated in 10 mM Tris-HCl, 1 mM EDTA, 6% trehalose, pH 8.0.
  • Approximate molecular weight 44 kDa.
  • Purity ≥90% as determined by reducing SDS-PAGE.
  • Store frozen; aliquot and keep at -20°C or -80°C for extended storage.

Catalog No. 50-001-77788 Supplier Medchemexpress LLC Supplier No. HYP700253100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Recombinant human desert hedgehog (DHH) protein supplied as a lyophilized preparation with N-terminal His and GST tags and a C-terminal Myc tag. Expressed in E. coli, the construct corresponds to amino acids G51-K158 of human DHH (accession O43323) and is intended for research on hedgehog signaling and gonadal development.

  • Recombinant human DHH with His-GST and Myc tags.
  • Expressed in E. coli and lyophilized for stability.
  • Formulated in 10 mM Tris-HCl, 1 mM EDTA, 6% trehalose, pH 8.0.
  • Approximate molecular weight 44 kDa.
  • Purity ≥90% as determined by reducing SDS-PAGE.
  • Store frozen; aliquot and keep at -20°C or -80°C for extended storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.