Learn More
TARGETMOL CHEMICALS INC DHODH-IN-1 5MG
SDP
Catalog No. 502581598
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. DHODH-IN-1 is a potent dihydroorotate dehydrogenase (DHODH) inhibitor with IC50 of 25 nM. DHODH-IN-1 is an inhibitor of the pyrimidine biosynthetic pathway. purity: 99%
Catalog No. 50-258-1598 Supplier TARGETMOL CHEMICALS INC Supplier No. T110195MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. DHODH-IN-1 is a potent dihydroorotate dehydrogenase (DHODH) inhibitor with IC50 of 25 nM. DHODH-IN-1 is an inhibitor of the pyrimidine biosynthetic pathway. purity: 99%