Learn More
Cayman Chemical DHODH-IN 11 10mg
SDP
Catalog No. 502913190
Encompass_Preferred
A derivative of leflunomide; does not inhibit DHODH (IC50 = >100 µM)
Catalog No. 50-291-3190 Supplier Cayman Chemical Supplier No. 3615910 MG
A derivative of leflunomide; does not inhibit DHODH (IC50 = >100 µM)