Learn More
TARGETMOL CHEMICALS INC DHODH-IN-13 5MG
SDP
Catalog No. 502581596
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. DHODH-IN-13 is an inhibitor of DHODH with IC50 of 4.3 uM for rat liver. DHODH-IN-13 can be used in studies about rheumatoid arthritis. purity: 99%
Catalog No. 50-258-1596 Supplier TARGETMOL CHEMICALS INC Supplier No. T110225MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. DHODH-IN-13 is an inhibitor of DHODH with IC50 of 4.3 uM for rat liver. DHODH-IN-13 can be used in studies about rheumatoid arthritis. purity: 99%