Learn More
TARGETMOL CHEMICALS INC DHODH-IN-15 5MG
SDP
Catalog No. 502581594
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. DHODH-IN-15 is the hydroxyfuran analogue of A771726. DHODH-IN-15 is a dihydroorotate dehydrogenase (DHODH) inhibitor with an IC50 of 11 uM for rat liver DHODH. DHODH-IN-15 can be used for rheumatoid arthritis. purity: 97%
Catalog No. 50-258-1594 Supplier TARGETMOL CHEMICALS INC Supplier No. T110245MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. DHODH-IN-15 is the hydroxyfuran analogue of A771726. DHODH-IN-15 is a dihydroorotate dehydrogenase (DHODH) inhibitor with an IC50 of 11 uM for rat liver DHODH. DHODH-IN-15 can be used for rheumatoid arthritis. purity: 97%