Learn More
Medchemexpress LLC HY-139189 5mg , DHODH-IN-16 CAS:2511248-11-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1391895MG
Encompass_Preferred
HY-139189 5mg DHODH-IN-16 CAS: 2511248-11-4 DHODH-IN-16 is a potent dihydroorotate dehydrogenase (DHODH) inhibitor with an IC50 of 0.396 nM for human DHODH. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1817