Learn More
Medchemexpress LLC DHODH-IN-17 | 16344-26-6 | 99.9% | 10 MG
SDP
Catalog No. 5000338315
Encompass_Preferred
DHODH-IN-17 is a 2-anilino nicotinic acid and a human DHODH inhibitor with an IC50 value of 0.40 μM. It is intended for research purposes, specifically in the study of acute myeloid leukemia (AML). Its molecular weight is 248.67, with a chemical formula of C12H9ClN2O2, and it appears as an off-white to gray solid.
  • Inhibits human DHODH with an IC50 of 0.4 μM.
  • Induces differentiation in bone marrow cells (EC50 of 3 μM).
  • Induces differentiation in THP-1 cells (EC50 of 10.7 μM).
  • Induces differentiation in U-937 cells (EC50 of 6.1 μM).
Catalog No. 50-003-38315 Supplier Medchemexpress LLC Supplier No. HY12806810MG
DHODH-IN-17 is a 2-anilino nicotinic acid and a human DHODH inhibitor with an IC50 value of 0.40 μM. It is intended for research purposes, specifically in the study of acute myeloid leukemia (AML). Its molecular weight is 248.67, with a chemical formula of C12H9ClN2O2, and it appears as an off-white to gray solid.
  • Inhibits human DHODH with an IC50 of 0.4 μM.
  • Induces differentiation in bone marrow cells (EC50 of 3 μM).
  • Induces differentiation in THP-1 cells (EC50 of 10.7 μM).
  • Induces differentiation in U-937 cells (EC50 of 6.1 μM).