Learn More
TARGETMOL CHEMICALS INC DHODH-IN-4 5MG
SDP
Catalog No. 502581592
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. DHODH-IN-4 is a human and Plasmodium falciparum dihydroorotate dehydrogenase (DHODH) inhibitor. The IC50 values of PfDHODH and HsDHODH are 4 uM and 0.18 uM respectively. DHODH-IN-4 has antimalarial activity. purity: 98%
Catalog No. 50-258-1592 Supplier TARGETMOL CHEMICALS INC Supplier No. T110275MG
Add to Cart
Add to Cart
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. DHODH-IN-4 is a human and Plasmodium falciparum dihydroorotate dehydrogenase (DHODH) inhibitor. The IC50 values of PfDHODH and HsDHODH are 4 uM and 0.18 uM respectively. DHODH-IN-4 has antimalarial activity. purity: 98%