Learn More
AdipoGen DHPDS
SDP
Supplier:  AdipoGen CDXD0078M100
Encompass_Preferred
Chemical. CAS 61255-63-8. Formula C16H8Na2O8S2. MW 438.34. Synthetic. Highly water soluble ratiometric pH indicator for the physiological range.
Catalog No. 50-256-1258