Learn More
Medchemexpress LLC DHPDS disodium salt | 61255-63-8 | 98.0% | 5 MG
SDP
Catalog No. 5000410535
Encompass_Preferred
DHPDS disodium salt is a pH-sensitive fluorescent dye utilized in research to measure intracellular pH (pHi) by observing the surface fluorescence of an isolated perfused rat liver. This product is intended for research use only.
  • pH-sensitive fluorescent dye
  • Used for measuring intracellular pH (pHi)
  • Light yellow to green yellow solid
Catalog No. 50-004-10535 Supplier Medchemexpress LLC Supplier No. HYD00755MG
DHPDS disodium salt is a pH-sensitive fluorescent dye utilized in research to measure intracellular pH (pHi) by observing the surface fluorescence of an isolated perfused rat liver. This product is intended for research use only.
  • pH-sensitive fluorescent dye
  • Used for measuring intracellular pH (pHi)
  • Light yellow to green yellow solid