Learn More
Medchemexpress LLC HY-12598A 5mg Medchemexpress, DHPG CAS:146255-66-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY12598A5MG
Encompass_Preferred
Medchemexpress, HY-12598A 5mg DHPG CAS:146255-66-5 DHPG ((RS)-3,5-DHPG) is an amino acid, which acts as a selective and potent agonist of group I mGluR (mGluR 1 and mGluR 5), shows no effect on Group II or Group III mGluRs. DHPG ((RS)-3,5-DHPG) is also an effective antagonist of mGluRs linked to phospholipase D. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9713