Learn More
Medchemexpress LLC (DHQD)2PHAL | 140853-10-7 | 778.98 | 10 G
SDP
Catalog No. 5000447842
Encompass_Preferred
(DHQD)2PHAL is a dimer that can be used as a ligand for the Sharpless asymmetric dihydroxylation reaction. It is a white to off-white solid with a molecular formula of C48H54N6O4.
  • Solid appearance
  • White to off-white color
  • Molecular formula: C48H54N6O4
  • Used as a ligand for Sharpless asymmetric dihydroxylation reaction
Catalog No. 50-004-47842 Supplier Medchemexpress LLC Supplier No. HYW00803310G
(DHQD)2PHAL is a dimer that can be used as a ligand for the Sharpless asymmetric dihydroxylation reaction. It is a white to off-white solid with a molecular formula of C48H54N6O4.
  • Solid appearance
  • White to off-white color
  • Molecular formula: C48H54N6O4
  • Used as a ligand for Sharpless asymmetric dihydroxylation reaction