Learn More
Aviva Systems Biology Corp DHRS2 Antibody - N-terminal region (ARP42258_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42258_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DHRS2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human DHRS2. Alternate names: HEP27, SDR25C1ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-284-988