Learn More
Aviva Systems Biology Corp Dhrs3 Antibody - middle region (ARP46495_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46495_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Dhrs3. It was validated on Western Blot. . Alternate names: Rsdr1, retSDR1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-285-261