Learn More
Aviva Systems Biology Corp DHRS4 Antibody - N-terminal region (ARP62272_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62272_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DHRS4. It was validated on Western Blot. . Alternate names: FLJ11008, NRDR, SCAD-SRL, SDR-SRL, SDR25C1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-287-641