Learn More
Aviva Systems Biology Corp Dhrs7 Antibody - N-terminal region (ARP47211_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47211_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Dhrs7. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: 2310016E22Rik, 5730564L20Rik, AW061210, Retdsr4, Retsdr4ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-285-289