Learn More
Aviva Systems Biology Corp DHRS9 Antibody - middle region (ARP53475_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53475_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DHRS9. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human DHRS9. Alternate names: 3alpha-HSD, RDH15, RDHL, RETSDR8, RDHTBE, SDR9C4, 3ALPHA-HSDApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61034