Learn More
Aviva Systems Biology Corp DHRSX Antibody - C-terminal region (ARP31650_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31650_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DHRSX. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human DHRSX. Alternate names: CXorf11, DHRS5X, DHRS5Y, DHRSXY, DHRSY, SDR46C1ApplicationWB. Species reactivity: Dog, Guinea Pig, Human, Rat
Catalog No. 50-014-54295