Learn More
Aviva Systems Biology Corp DHRSX Antibody - N-terminal region (ARP39847_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39847_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DHRSX. It was validated on Western Blot. . Alternate names: CXorf11, DHRS5X, DHRS5Y, DHRSXY, DHRSY, SDR46C1ApplicationWB. Species reactivity: Cow, Human, Rat, Yeast
Catalog No. 50-284-838