Learn More
Aviva Systems Biology Corp DHX15 Antibody - C-terminal region (ARP36347_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36347_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DHX15. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human DHX15. Alternate names: DBP1, DDX15, HRH2, PRP43, PRPF43, PrPp43pApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56122