Learn More
Aviva Systems Biology Corp DHX30 Antibody - N-terminal region (ARP36425_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36425_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DHX30. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human DHX30. Alternate names: DDX30, FLJ11214, KIAA0890, Ret-CoR, RETCORApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-56150