Learn More
Aviva Systems Biology Corp DHX32 Antibody - N-terminal region (ARP36451_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36451_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DHX32. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human DHX32. Alternate names: DDX32, DHLP1, FLJ10694, FLJ10889ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-53047