Learn More
Aviva Systems Biology Corp DHX37 Antibody - N-terminal region (ARP36514_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36514_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DHX37. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human DHX37. Alternate names: DDX37, FLJ41974, KIAA1517, MGC2695, MGC4322, MGC46245ApplicationWB. Species reactivity: Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-56178