Learn More
Aviva Systems Biology Corp DHX57 Antibody - C-terminal region (ARP36544_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36544_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DHX57. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human DHX57. Alternate names: DDX57, FLJ32861ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-284-500