Learn More
Aviva Systems Biology Corp DHX58 Antibody - N-terminal region (ARP36500_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36500_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DHX58. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human DHX58. Alternate names: D11LGP2, D11lgp2e, LGP2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-56176