Learn More
Aviva Systems Biology Corp DHX9 Antibody - N-terminal region (ARP36343_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36343_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DHX9. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human DHX9. Alternate names: DDX9, LKP, NDH II, NDHII, RHA, NDH2ApplicationIHC, WB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-56120