Learn More
Strem, An Ascensus Company CAS# 131266-79-0. 1g. Di-1-adamantylphosphine oxide, min. 90%. MFCD00185311
SDP
Supplier:  Strem, An Ascensus Company 1509561G
Encompass
CAS# 131266-79-0. 1g. Di-1-adamantylphosphine oxide, min. 90%. MFCD00185311. Molecular Weight: 318.43. Molecular Formula: (C10H15)2P(O)H. Color/form: white pwdr. Strem# 15-0956. http://www.strem.com/catalog/v/15-0956/
Catalog No. 50-901-09983