Learn More
Biotium Di-12-ANEPPQ
Supplier:  Biotium 61015
Encompass_Preferred
Di-12-ANEPPQ is a fast-responding membrane potential dye. It is more hydrophobic than Di-8-ANEPPQ and is useful for potential-sensitive retrograde labeling of neurons. Properties: Orange red solid soluble in DMF or DMSO; Store at -20°C and protect from light, especially in solution; C46H75Br2N3; MW: 827.4
Catalog No. 50-196-4736