Learn More
Medchemexpress LLC DI-1859 | 2247061-09-0 | 99.6% | C30H45N5O3S | 1 ML
SDP
Catalog No. 5000355924
Encompass_Preferred
DI-1859 is a potent, selective, and covalent inhibitor of DCN1. It inhibits neddylation of cullin 3 in cells at low nanomolar concentrations. DI-1859 increases NRF2 protein, a CRL3 substrate, in mouse liver and protects mice from acetaminophen-induced liver damage.
  • Potent, selective, and covalent inhibitor of DCN1
  • Inhibits neddylation of cullin 3 in cells
  • Increases NRF2 protein in mouse liver
  • Protects mice from acetaminophen-induced liver damage
  • For research use only
Catalog No. 50-003-55924 Supplier Medchemexpress LLC Supplier No. HY14573310MM/1ML
DI-1859 is a potent, selective, and covalent inhibitor of DCN1. It inhibits neddylation of cullin 3 in cells at low nanomolar concentrations. DI-1859 increases NRF2 protein, a CRL3 substrate, in mouse liver and protects mice from acetaminophen-induced liver damage.
  • Potent, selective, and covalent inhibitor of DCN1
  • Inhibits neddylation of cullin 3 in cells
  • Increases NRF2 protein in mouse liver
  • Protects mice from acetaminophen-induced liver damage
  • For research use only