Learn More
Biotium Di-2-ANEPEQ
Catalog No. 501964734
Encompass_Preferred
Di-2-ANEPEQ, also called JPW1114, is a fast-responding membrane potential dye. It is highly water-soluble and is usually introduced into cells by microinjection. The dye can also be applied to brain tissues topically.Properties: Orange red solid soluble in H2O, ethanol or DMSO; Store at -20°C and protect from light, especially in solution; C26H35Br2N3; MW: 547.12
Catalog No. 50-196-4734 Supplier Biotium Supplier No. 61013
Di-2-ANEPEQ, also called JPW1114, is a fast-responding membrane potential dye. It is highly water-soluble and is usually introduced into cells by microinjection. The dye can also be applied to brain tissues topically.Properties: Orange red solid soluble in H2O, ethanol or DMSO; Store at -20°C and protect from light, especially in solution; C26H35Br2N3; MW: 547.12